Learn More
CellSensor™ IL1-bla ECV304 Cell Line
CellSensor™ IL-1-bla ECV304 cell line is an adherent human urinary bladder cell line used to measure the (IL-1) signaling pathway via transcriptional regulation of the beta-lactamase gene.
Brand: CellSensor™ K1645
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- IL-1 cytokine cell signaling is a critical pathway involved in immune and inflammatory responses.
- This cell line was constructed using GenomeScreen™ technology utilizing a promoterless beta-lactamase reporter gene randomly integrated into the genome of the IL-1-bla ECV304 cell line.
- Sequential rounds of sorting were performed to isolate cell clones containing tagged genes induced by IL-1.
- A dose response assay was run with this cell line using IL-1 as the agonist.
Specifications
Store in liquid nitrogen immediately upon receipt, or thaw for immediate use | |
Cytokines, Signaling Pathway | |
1 Vial | |
Dividing Cells |