missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-9R Antibody (RM0115-12F43) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£153.00 - £426.00
Specifications
| Antigen | IL-9R |
|---|---|
| Clone | RM0115-12F43 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18233592
|
Novus Biologicals
NBP1-22493-0.025MG |
0.025 mg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18225900
|
Novus Biologicals
NBP1-22493 |
0.1 mg |
£426.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-9R Monoclonal specifically detects IL-9R in Mouse samples. It is validated for Western Blot.Specifications
| IL-9R | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| 3581 | |
| Mouse recombinant protein of the extracellular domain of IL-9 R | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-9 R. |
| RM0115-12F43 | |
| Western Blot 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
| IL9R | |
| IgG2 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title