missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-9R Antibody (RM0115-12F43) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22493-0.025MG
This item is not returnable.
View return policy
Description
IL-9R Monoclonal specifically detects IL-9R in Mouse samples. It is validated for Western Blot.
Specifications
| IL-9R | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000 | |
| CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
| Rat | |
| Protein G purified | |
| RUO | |
| 3581 | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
| Western Blot | |
| RM0115-12F43 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| IL9R | |
| Mouse recombinant protein of the extracellular domain of IL-9 R | |
| 0.025 mg | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-9 R. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction