missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-9R Antibody (4A11H2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£184.00 - £470.00
Specifications
| Antigen | IL-9R |
|---|---|
| Clone | 4A11H2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18253851
|
Novus Biologicals
NBP2-61701 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647496
|
Novus Biologicals
NBP2-61701-0.025ml |
0.025 mL |
£184.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-9R Monoclonal specifically detects IL-9R in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IL-9R | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
| IL9R | |
| IgG1 | |
| Protein G purified |
| 4A11H2 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| Human, Rat | |
| 3581 | |
| Purified recombinant fragment of human CD129 (AA: extra 41-270) expressed in E. Coli. | |
| Primary | |
| 57.1 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title