missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-9R Antibody (4A11H2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61701
This item is not returnable.
View return policy
Description
IL-9R Monoclonal specifically detects IL-9R in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-9R | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
| Mouse | |
| 57.1 kDa | |
| 0.1 mL | |
| 3581 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4A11H2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| IL9R | |
| Purified recombinant fragment of human CD129 (AA: extra 41-270) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction