Learn More
IL-9 Monoclonal Antibody (D9302C12), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
£7770.00 - £12950.00
Specifications
| Antigen | IL-9 |
|---|---|
| Clone | D9302C12 |
| Concentration | 1 mg/mL |
| Applications | Functional Assay, Neutralization |
| Classification | Monoclonal |
Description
This monoclonal D9302C12 antibody reacts with and inhibits the bioactivity of mouse IL-9. IL-9 is a 14 kDa cytokine originally named P40 and identified by its proliferative effects on T cell populations. The receptor, which is a heterodimer of the gamma chain portion of the IL-2 receptor and the IL-9R chain, activates Jak/STAT signaling pathways upon binding its ligand. Since the discovery of IL-9, numerous other functions have been observed. It induces Th17 and Treg differentiation in CD4+ T cells, IgE production in B cells, and the differentiation and proliferation of mast cells. IL-9 expression was initially observed in Th2 cells, but has since been found in Th17, eosinophil, and mast cells. Th9 cells, a newly discovered subset of CD4+ T cells, are characterized by the secretion of large amounts of IL-9 and IL-10. Th9 development is induced by stimulation of undifferentiated CD4+ with IL-4 and TGF beta. Th2 cells can also be driven towards a Th9 phenotype in the presence of TGF beta.
IL-9 Monoclonal antibody specifically detects IL-9 in Mouse samples. It is validated for Functional Assay, NeutralizationSpecifications
| IL-9 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; Il9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor p40 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| D9302C12 | |
| Functional Assay, Neutralization | |
| Functional Grade | |
| Armenian Hamster | |
| Mouse | |
| P15247 | |
| 16198 | |
| Primary | |
| 4° C | |
| Il9 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.