missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-6 Receptor Recombinant Rabbit Monoclonal Antibody (9S8Q5)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£147.00 - £363.00

Specifications

Antigen IL-6 Receptor
Clone 9S8Q5
Concentration 1.07 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30284397
View Documents
Invitrogen™
MA557953
20 μL
£147.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30282522
GREENER_CHOICE
View Documents
Invitrogen™
MA557954
100 μL
£363.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: LVLGSCRALE VANGTVTSLP GATVTLICPG KEAAGNVTIH WVYSGSQNRE WTTTGNTLVL RDVQLSDTGD YLCSLNDHLV GTVPLLVDVP PEEPKLSCFR KNPLVNAICE WRPSSTPSPT TKAVLFAKKI NTTNGKSDFQ VPCQYSQQLK SFSCQVEILE GDKVYHIVSL CVANSVGSKS SHNEAFHSLK MVQPDPPANL VVSAIPGRPR WLKVSWQHPE TWDPSYYLLQ FQLRYRPVWS KEFTVLLLPV AQYQCVIHDA LRGVKHVVQV RGKEELDLGQ WSEWSPEVTG TPWIAEPRTT PAGILWNPTQ VSVEDSANHE DQYESSTEAT SVLAPVQE.

Interleukin 6 receptor (IL6R), also known as CD126 (Cluster of Differentiation 126), is a type I cytokine receptor. This gene encodes a subunit of the interleukin 6 (IL6) receptor complex. Interleukin 6 is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in the immune response. The IL6 receptor is a protein complex consisting of this protein and interleukin 6 signal transducer (IL6ST/GP130/IL6-beta), a receptor subunit also shared by many other cytokines. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer. Alternatively spliced transcript variants encoding distinct isoforms have been reported. A pseudogene of this gene is found on chromosome 9.
Specifications

Specifications

IL-6 Receptor
1.07 mg/mL
ELISA, Flow Cytometry, Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P22272
16194
Recombinant Protein (or fragment)
Antibody
9S8Q5
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Mouse
Il6ra
CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; IL6R; IL-6R; IL-6R 1; IL-6R subunit alpha; IL6R1; IL-6R-1; Il6ra; IL-6RA; IL-6R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991
Il6ra
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.