Learn More
Invitrogen™ IL-6 Receptor Recombinant Rabbit Monoclonal Antibody (9S8Q5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557954
Description
Immunogen sequence: LVLGSCRALE VANGTVTSLP GATVTLICPG KEAAGNVTIH WVYSGSQNRE WTTTGNTLVL RDVQLSDTGD YLCSLNDHLV GTVPLLVDVP PEEPKLSCFR KNPLVNAICE WRPSSTPSPT TKAVLFAKKI NTTNGKSDFQ VPCQYSQQLK SFSCQVEILE GDKVYHIVSL CVANSVGSKS SHNEAFHSLK MVQPDPPANL VVSAIPGRPR WLKVSWQHPE TWDPSYYLLQ FQLRYRPVWS KEFTVLLLPV AQYQCVIHDA LRGVKHVVQV RGKEELDLGQ WSEWSPEVTG TPWIAEPRTT PAGILWNPTQ VSVEDSANHE DQYESSTEAT SVLAPVQE.
Interleukin 6 receptor (IL6R), also known as CD126 (Cluster of Differentiation 126), is a type I cytokine receptor. This gene encodes a subunit of the interleukin 6 (IL6) receptor complex. Interleukin 6 is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in the immune response. The IL6 receptor is a protein complex consisting of this protein and interleukin 6 signal transducer (IL6ST/GP130/IL6-beta), a receptor subunit also shared by many other cytokines. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer. Alternatively spliced transcript variants encoding distinct isoforms have been reported. A pseudogene of this gene is found on chromosome 9.
Specifications
| IL-6 Receptor | |
| Recombinant Monoclonal | |
| 1.07 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P22272 | |
| Il6ra | |
| Recombinant Protein (or fragment) | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Flow Cytometry, Western Blot | |
| 9S8Q5 | |
| Unconjugated | |
| Il6ra | |
| CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; IL6R; IL-6R; IL-6R 1; IL-6R subunit alpha; IL6R1; IL-6R-1; Il6ra; IL-6RA; IL-6R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 16194 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.