Learn More
IL-5 Rat anti-Human, PE, Clone: JES1-39D10, BD
Rat Monoclonal Antibody
£90.74 - £312.61
Specifications
| Antigen | IL-5 |
|---|---|
| Clone | JES1-39D10 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15897728
|
BD Biosciences
554489 |
0.1 mg |
£326.00 £312.61 / Each Save £13.39 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15889708
|
BD Biosciences
559332 |
100 Tests |
£94.20 £90.74 / Each Save £3.46 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The JES1-39D10 antibody reacts with human interleukin-5 (IL-5). The immunogen used to generate the JES1-39D10 hybridoma was COS-expressed recombinant human IL-5. This is a neutralizing antibody. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Intracellular Staining
Specifications
| IL-5 | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| Immunology | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2a | |
| Affinity Purified |
| JES1-39D10 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Human | |
| Recombinant human IL-5 | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.