Learn More
IL-5 Rat anti-Human, PE, Clone: JES1-39D10, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 559332
Description
The JES1-39D10 antibody reacts with human interleukin-5 (IL-5). The immunogen used to generate the JES1-39D10 hybridoma was COS-expressed recombinant human IL-5. This is a neutralizing antibody. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Intracellular Staining
Specifications
| IL-5 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant human IL-5 | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| JES1-39D10 | |
| PE | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.