missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£224.00 - £654.00
Specifications
| Antigen | IL-5 |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18646698
|
Novus Biologicals
NBP3-11324 |
0.1 mg |
£654.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18684667
|
Novus Biologicals
NBP3-11324-0.025mg |
0.025 mg |
£224.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-5 Polyclonal antibody specifically detects IL-5 in Bovine samples. It is validated for Western Blot, ELISASpecifications
| IL-5 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine | |
| B-cell differentiation factor I, EDF, Eosinophil differentiation factor, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRFB cell differentiation factor I | |
| Recombinant bovine IL-5 | |
| Primary | |
| Store at 4°C. Do not freeze. |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology | |
| PBS | |
| 3567 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title