missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-11324-0.025mg
This item is not returnable.
View return policy
Description
IL-5 Polyclonal antibody specifically detects IL-5 in Bovine samples. It is validated for Western Blot, ELISA
Specifications
| IL-5 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| B-cell differentiation factor I, EDF, Eosinophil differentiation factor, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRFB cell differentiation factor I | |
| Recombinant bovine IL-5 | |
| 0.025 mg | |
| Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology | |
| 3567 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction