missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4 Rat, APC, Clone: 11B11, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB81-APC-100ug
Additional Details : Weight : 0.01000kg
Specifications
IL-4 | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Il4 | |
Protein A/G Purified | |
RUO | |
16189 | |
IgG1 κ |
Flow Cytometry | |
11B11 | |
APC | |
NP_067258 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |