missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4 Rat, APC, Clone: 11B11, enQuireBio™
Rat Monoclonal Antibody
£71.60 - £237.00
Specifications
Antigen | IL-4 |
---|---|
Clone | 11B11 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962538
|
EnquireBio
QAB81-APC-25UG |
25 μg |
£71.60
25µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952538
|
EnquireBio
QAB81-APC-100UG |
100 μg |
£237.00
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IL-4 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
16189 | |
IgG1 κ | |
Protein A/G Purified |
11B11 | |
Flow Cytometry | |
APC | |
RUO | |
NP_067258 | |
Il4 | |
Primary | |
Store at 2-4°C. |