missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4 Antibody (V17P1B4*B12) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £378.00
Specifications
| Antigen | IL-4 |
|---|---|
| Clone | V17P1B4*B12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18222642
|
Novus Biologicals
NBP2-61126 |
0.1 mg |
£378.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643287
|
Novus Biologicals
NBP2-61126-0.025mg |
0.025 mg |
£150.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-4 Monoclonal specifically detects IL-4 in Fish samples. It is validated for Western Blot, ELISA.Specifications
| IL-4 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Fish | |
| B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| IL4 | |
| IgG | |
| Protein A purified |
| V17P1B4*B12 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| PBS with 0.02% Sodium Azide | |
| 3565 | |
| Ovalbumin-conjugated synthetic peptide- AQAIYSRPHP | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title