missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4 Antibody (V17P1B4*B12) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61126
This item is not returnable.
View return policy
Description
IL-4 Monoclonal specifically detects IL-4 in Fish samples. It is validated for Western Blot, ELISA.
Specifications
| IL-4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
| B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3565 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| V17P1B4*B12 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| IL4 | |
| Ovalbumin-conjugated synthetic peptide- AQAIYSRPHP | |
| 0.1 mg | |
| Primary | |
| Fish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction