missing translation for 'onlineSavingsMsg'
Learn More

IL-3R alpha/CD123, Rabbit anti-Human, Clone: IL3RA/2947R, Novus Biologicals™

Rabbit Monoclonal Antibody

£246.00 - £431.00

Specifications

Antigen IL-3R alpha/CD123
Clone IL3RA/2947R
Concentration 0.2 mg/mL
Dilution ELISA, Immunohistochemistry 0.25-0.5 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml, Protein Array
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18256537
View Documents
Novus Biologicals
NBP2-79717-20UG
20 μg
£246.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18211798
View Documents
Novus Biologicals
NBP2-79717-100UG
100 μg
£431.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-3R alpha/CD123 Monoclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

IL-3R alpha/CD123
0.2 mg/mL
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Rabbit
B Cell Development and Differentiation Markers, Cytokine Research, Oncogenes, Phospho Specific, Protein Kinase
10 mM PBS with 0.05% BSA
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
IL3RA
IgG
Protein A or G purified
IL3RA/2947R
ELISA, Immunohistochemistry 0.25-0.5 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml, Protein Array
Monoclonal
Purified
RUO
Human
P26951, P26951
3563
Recombinant fragment of human IL-3R alpha/CD123 protein (around aa 26-171) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.