missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-3R alpha/CD123, Rabbit anti-Human, Clone: IL3RA/2947R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-79717-100UG
This item is not returnable.
View return policy
Description
IL-3R alpha/CD123 Monoclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| IL-3R alpha/CD123 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry 0.25-0.5 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml, Protein Array | |
| P26951, P26951 | |
| IL3RA | |
| Recombinant fragment of human IL-3R alpha/CD123 protein (around aa 26-171) (exact sequence is proprietary) | |
| 100 μg | |
| B Cell Development and Differentiation Markers, Cytokine Research, Oncogenes, Phospho Specific, Protein Kinase | |
| 3563 | |
| Store at 4°C. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| IL3RA/2947R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction