Learn More
IL-36 Beta/IL-1F8 Mouse anti-Human, Unconjugated, Clone: 10D12B1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | IL-36 Beta/IL-1F8 |
|---|---|
| Clone | 10D12B1 |
| Concentration | 940 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16827365
|
Proteintech
60290-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16817365
|
Proteintech
60290-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The IL-36 Beta/IL-1F8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-36 Beta/IL-1F8. This antibody recognizes human antigen. The IL-36 Beta/IL-1F8 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| IL-36 Beta/IL-1F8 | |
| 940 μg/mL | |
| Monoclonal | |
| Mouse | |
| 27177 | |
| Primary |
| 10D12B1 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.