Learn More
IL-36 Beta/IL-1F8 Mouse anti-Human, Unconjugated, Clone: 10D12B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60290-1-IG-150UL
Description
The IL-36 Beta/IL-1F8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-36 Beta/IL-1F8. This antibody recognizes human antigen. The IL-36 Beta/IL-1F8 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| IL-36 Beta/IL-1F8 | |
| Monoclonal | |
| 940 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 10D12B1 | |
| Unconjugated | |
| 150 μL | |
| 27177 | |
| IgG1 |