missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-29/IFN-lambda 1 Mouse anti-Human, Clone: B-S29, Novus Biologicals™
Mouse Monoclonal Antibody
£305.00 - £571.00
Specifications
| Antigen | IL-29/IFN-lambda 1 |
|---|---|
| Clone | B-S29 |
| Concentration | 1 mg/ml |
| Dilution | ELISA |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18364586
|
Novus Biologicals
NBP3-18106-200UL |
200 μg |
£305.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18315844
|
Novus Biologicals
NBP3-18106-500UL |
500 μg |
£571.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-29/IFN-lambda 1 Monoclonal antibody specifically detects IL-29/IFN-lambda 1 in Human samples. It is validated for ELISASpecifications
| IL-29/IFN-lambda 1 | |
| 1 mg/ml | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins | |
| PBS. Sterile-filtered through 0.22 um. | |
| 282618 | |
| IgG2b κ | |
| Affinity purified |
| B-S29 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cytokine Zcyto21, IFNL1, IFN-lambda-1, IL-29, interferon lambda-1, interferon, lambda 1, interleukin 29 (interferon, lambda 1), interleukin-29, ZCYTO21 | |
| Recombinant human IL-29/IFN-lambda 1. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title