missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-29/IFN-lambda 1 Mouse anti-Human, Clone: B-S29, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18106-500UL
This item is not returnable.
View return policy
Description
IL-29/IFN-lambda 1 Monoclonal antibody specifically detects IL-29/IFN-lambda 1 in Human samples. It is validated for ELISA
Specifications
| IL-29/IFN-lambda 1 | |
| Monoclonal | |
| 1 mg/ml | |
| ELISA | |
| cytokine Zcyto21, IFNL1, IFN-lambda-1, IL-29, interferon lambda-1, interferon, lambda 1, interleukin 29 (interferon, lambda 1), interleukin-29, ZCYTO21 | |
| Recombinant human IL-29/IFN-lambda 1. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| 500 μg | |
| Biologically Active Proteins | |
| 282618 | |
| Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| ELISA | |
| B-S29 | |
| Unconjugated | |
| PBS. Sterile-filtered through 0.22 um. | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction