missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-22R alpha 1 Goat anti-Human, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
£155.00
Specifications
| Antigen | IL-22R alpha 1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry |
| Classification | Polyclonal |
Description
IL-22R alpha 1 Polyclonal specifically detects IL-22R alpha 1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IL-22R alpha 1 | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Polyclonal | |
| Goat | |
| Human | |
| CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R1, IL-22RA1, IL-22R-alpha-1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
| IL22RA1 | |
| IgG | |
| Affinity Purified | |
| IL22 Receptor |
| 1.0 mg/mL | |
| Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA with 0.1% Sodium Azide | |
| 58985 | |
| Synthetic peptide: DSGPEGTPDTVYSIEYKTYGERDWVAKK, corresponding to N terminal amino acids 42-70 of Human IL22 Receptor. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title