missing translation for 'onlineSavingsMsg'
Learn More

IL-22R alpha 1 Goat anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18260105 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18260105 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18260105 Supplier Novus Biologicals Supplier No. NB1007400.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-22R alpha 1 Polyclonal specifically detects IL-22R alpha 1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-22R alpha 1
Applications Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA with 0.1% Sodium Azide
Gene Alias CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R1, IL-22RA1, IL-22R-alpha-1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11
Gene Symbols IL22RA1
Host Species Goat
Immunogen Synthetic peptide: DSGPEGTPDTVYSIEYKTYGERDWVAKK, corresponding to N terminal amino acids 42-70 of Human IL22 Receptor.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58985
Test Specificity IL22 Receptor
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.