missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-20R alpha Antibody (MM0394-9F40) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£153.00 - £436.00
Specifications
| Antigen | IL-20R alpha |
|---|---|
| Clone | MM0394-9F40 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18055787
|
Novus Biologicals
NBP2-11695-0.025MG |
0.025 mg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18027705
|
Novus Biologicals
NBP2-11695 |
0.1 mg |
£436.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-20R alpha Monoclonal specifically detects IL-20R alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| IL-20R alpha | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Human | |
| class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20R1IL-20 receptor subunit alpha, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7 | |
| IL20RA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0394-9F40 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q9UHF4 | |
| 53832 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title