missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-20R alpha Antibody (MM0394-9F40) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11695
This item is not returnable.
View return policy
Description
IL-20R alpha Monoclonal specifically detects IL-20R alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IL-20R alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:20-100, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:20-1:100 | |
| Q9UHF4 | |
| IL20RA | |
| Human recombinant IL-20R alpha (extracellular domain) | |
| 0.1 mg | |
| Primary | |
| Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MM0394-9F40 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20R1IL-20 receptor subunit alpha, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 53832 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction