Learn More
Invitrogen™ IL-2 Monoclonal Antibody (JES6-1A12), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£2250.00 - £12950.00
Specifications
| Antigen | IL-2 |
|---|---|
| Clone | JES6-1A12 |
| Concentration | 1 mg/mL |
| Classification | Monoclonal |
| Conjugate | Functional Grade |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17171452
|
Invitrogen™
16-7022-38 |
5 mg |
£2900.00
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15597796
|
Invitrogen™
16-7022-95 |
10 mg |
£2250.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17151452
|
Affymetrix eBioscience
16-7022-025 |
25 mg | |||||||
|
17161452
|
Affymetrix eBioscience
16-7022-050 |
50 mg | |||||||
Description
The JES6-1A12 antibody reacts with mouse interleukin-2 (IL-2). The JES6-1A12 antibody is a neutralizing antibody. Mouse IL-2 (also known as TCGF) is a 17 kDa factor produced mainly by activated CD4+ T cells. IL-2 induces cell cycle progression of resting cells in an antigen non-specific manner and allows clonal expansion of activated T cells. IL-2 also acts on activated B cells, monocytes, NK cells, LAK cells, and on oligodendroglial cells in vitro. In addition, IL-2 plays a role in hematopoiesis, tumor surveillance and anti-inflammatory reactions, and hence is a central regulator of the immune response. Non-glycosylated IL-2 is biologically active.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an μgcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
| IL-2 | |
| 1 mg/mL | |
| Functional Grade | |
| Rat | |
| Mouse | |
| P04351 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| JES6-1A12 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| 16183 | |
| Primary | |
| 4°C | |
| IL2 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.