Learn More
IL-2 Monoclonal Antibody (JES6-1A12), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-7022-050
Description
The JES6-1A12 antibody reacts with mouse interleukin-2 (IL-2). The JES6-1A12 antibody is a neutralizing antibody. Mouse IL-2 (also known as TCGF) is a 17 kDa factor produced mainly by activated CD4+ T cells. IL-2 induces cell cycle progression of resting cells in an antigen non-specific manner and allows clonal expansion of activated T cells. IL-2 also acts on activated B cells, monocytes, NK cells, LAK cells, and on oligodendroglial cells in vitro. In addition, IL-2 plays a role in hematopoiesis, tumor surveillance and anti-inflammatory reactions, and hence is a central regulator of the immune response. Non-glycosylated IL-2 is biologically active.
IL-2 Monoclonal antibody specifically detects IL-2 in Mouse samples. It is validated for ELISA, Functional Assay, Immunoprecipitation, Neutralization
Specifications
| IL-2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| P04351 | |
| Rat | |
| 50 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
| ELISA, Functional Assay, Immunoprecipitation, Neutralization | |
| JES6-1A12 | |
| Functional Grade | |
| IL2 | |
| Aldesleukin; cytokine; H-IL-2; il 2; Il2; IL-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; Interleukin2; interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; TCGF | |
| Affinity chromatography | |
| RUO | |
| 16183 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.