missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17E/IL-25 Antibody (RM0263-5J46) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£171.00 - £423.00
Specifications
| Antigen | IL-17E/IL-25 |
|---|---|
| Clone | RM0263-5J46 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18020587
|
Novus Biologicals
NBP2-11677-0.025MG |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18749553
|
Novus Biologicals
NBP2-11677 |
||||||||
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| IL-17E/IL-25 | |
| LYOPH | |
| Unconjugated | |
| Rat | |
| Mouse | |
| IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25 | |
| IL25 | |
| Primary |
| RM0263-5J46 | |
| Monoclonal | |
| Purified | |
| RUO | |
| NP_542767 | |
| 64806 | |
| IgG2 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title