missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17E/IL-25 Antibody (RM0263-5J46) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11677-0.025MG
This item is not returnable.
View return policy
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| IL-17E/IL-25 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:50-200, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| NP_542767 | |
| IL25 | |
| Produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of IL-17E. | |
| 0.025 mg | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| RM0263-5J46 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 64806 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction