missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17E/IL-25 Antibody (68C1039.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
£177.00 - £407.00
Specifications
| Antigen | IL-17E/IL-25 |
|---|---|
| Clone | 68C1039.2 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:250, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 35359953), Immunohistochemistry-Paraffin reported in scientific literature (Terrier B et al) |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18705903
|
Novus Biologicals
NB100-56541SS |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18759763
|
Novus Biologicals
NB100-56541 |
0.1 mg |
£407.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IL-17E/IL-25 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25 | |
| IL25 | |
| IgG1 | |
| Protein G purified | |
| 20 kDa |
| 68C1039.2 | |
| Western Blot 1:100 - 1:250, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 35359953), Immunohistochemistry-Paraffin reported in scientific literature (Terrier B et al) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q9H293 | |
| 64806 | |
| Amino acids 115-132 of human IL-17E (isoform CRA_a) were used as immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title