missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17E/IL-25 Antibody (68C1039.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB100-56541
This item is not returnable.
View return policy
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-17E/IL-25 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:250, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 35359953), Immunohistochemistry-Paraffin reported in scientific literature (Terrier B et al) | |
| IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25 | |
| Mouse | |
| 20 kDa | |
| 0.1 mg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 68C1039.2 | |
| Unconjugated | |
| Q9H293 | |
| IL25 | |
| Amino acids 115-132 of human IL-17E (isoform CRA_a) were used as immunogen for this antibody. | |
| Protein G purified | |
| RUO | |
| 64806 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction