Learn More
IL-16 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 500-P06BT-1MG
Description
AA Sequence of recombinant protein: PDLNSSTDSA ASASAASDVS VESTAEATVC TVTLEKMSAG LGFSLEGGKG SLHGDKPLTI NRIFKGAASE QSETVQPGDE ILQLGGTAMQ GLTRFEAWNI IKALPDGPVT IVIRRKSLQS KETTAAGDS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-16. Anti-Human IL-16-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IL-16 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-16 (500-P06) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-16. Western Blot: To detect hIL-16 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIL-16 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P06BT-1MG will be provided as 2 x 500 μg
The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.
Specifications
| IL-16 | |
| Polyclonal | |
| Biotin | |
| IL16 | |
| FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3603 | |
| -20°C |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q14005 | |
| IL16 | |
| E.coli-derived Recombinant Human IL-16 (129 a.a.) | |
| 2 x 500μg | |
| Primary | |
| Human | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.