Learn More
IL-10 Rat anti-Human, APC, Clone: JES3-19F1, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 562036
Additional Details : Weight : 0.01000kg
Description
The JES3-19F1 antibody reacts with human interleukin-10 (IL-10). The immunogen used to generate the JES3-19F1 hybridoma was recombinant human IL-10 expressed in COS cells. This is a neutralizing antibody. This antibody also reacts with viral IL-10.
Specifications
IL-10 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant Human IL-10 | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
JES3-19F1 | |
APC | |
Rat | |
Affinity Purified | |
RUO | |
Human, Virus | |
IgG2a |
For Research Use Only.