missing translation for 'onlineSavingsMsg'
Learn More

IL-10 Rat anti-Human, APC, Clone: JES3-19F1, BD

Rat Monoclonal Antibody

£100.00 - £407.71

Specifications

Antigen IL-10
Clone JES3-19F1
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15891579
View Documents
BD Biosciences
562036
25 μg
ONLINE EXCLUSIVE PRICE
£104.00
£100.00 / Each
Save £4.00
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15817858
View Documents
BD Biosciences
554707
0.1 mg
ONLINE EXCLUSIVE PRICE
£424.00
£407.71 / Each
Save £16.29
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The JES3-19F1 antibody reacts with human interleukin-10 (IL-10). The immunogen used to generate the JES3-19F1 hybridoma was recombinant human IL-10 expressed in COS cells. This is a neutralizing antibody. This antibody also reacts with viral IL-10.

Specifications

Specifications

IL-10
0.2mg/mL
Monoclonal
Rat
Human, Virus
Recombinant Human IL-10
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
JES3-19F1
Flow Cytometry
APC
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG2a
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.