missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 Rabbit, anti-Rat, Clone: 1A4, Proteintech
Rabbit Recombinant Monoclonal Antibody
£158.43 - £442.11
Specifications
| Antigen | IL-10 |
|---|---|
| Clone | 1A4 |
| Concentration | 300 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
Description
The IL-10 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of rat IL-10. This antibody recognizes rat, mouse antigen. The IL-10 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IL-10 | |
| 300 μg/mL | |
| Liquid | |
| Rat, Mouse | |
| Il10, IL-10, IL10X, interleukin 10 | |
| Primary |
| 1A4 | |
| Western Blot, ELISA | |
| Rabbit | |
| 25325 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title