Learn More
IL-10 Rabbit, anti-Rat, Clone: 1A4, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82793-16-RR-20UL
Description
The IL-10 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of rat IL-10. This antibody recognizes rat, mouse antigen. The IL-10 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IL-10 | |
| 1A4 | |
| Il10, IL-10, IL10X, interleukin 10 | |
| 20 μL | |
| 25325 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Rat, Mouse | |
| 150 kDa | |
| IgG |