missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-10 Monoclonal Antibody (JES3-9D7)
Rat Monoclonal Antibody
£436.00 - £582.00
Specifications
| Antigen | IL-10 |
|---|---|
| Clone | JES3-9D7 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C |
| Applications | ELISA, Flow Cytometry, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
10008602
|
Invitrogen™
AHC0102 |
500 μg |
£436.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10241973
|
Invitrogen™
AHC0103 |
1 mg |
£582.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-10 Monoclonal Antibody for Flow, ELISA, Neu
IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.Specifications
| IL-10 | |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Neutralization | |
| Unconjugated | |
| Rat | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| P22301 | |
| 3586 | |
| Recombinant human Interleukin 10. | |
| Antibody |
| JES3-9D7 | |
| -20°C | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human | |
| IL10 | |
| CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF | |
| IL10 | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title