Learn More
Invitrogen™ IL-10 Monoclonal Antibody (JES3-9D7)
Rat Monoclonal Antibody
Brand:  Invitrogen™ AHC0103
 View more versions of this product
Product Code. 10241973
  • £582.00 / 1mg
For Research Use Only. All usage must comply with product instructions.
Description
Description
IL-10 Monoclonal Antibody for Flow, ELISA, Neu
IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-10
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
P22301
IL10
Recombinant human Interleukin 10.
1 mg
Primary
Human
Antibody
IgG1 κ
ELISA, Flow Cytometry, Neutralization
JES3-9D7
Unconjugated
IL10
CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Rat
Purified
RUO
3586
-20°C