missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IL-1Rrp2/IL-1R6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £452.00
Specifications
| Antigen | IL-1Rrp2/IL-1R6 |
|---|---|
| Clone | 131011 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18725679
![]() |
R&D Systems
MAB573-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18190511
|
R&D Systems
MAB573 |
500 μg |
£452.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-1Rrp2/IL-1R6 Monoclonal specifically detects IL-1Rrp2/IL-1R6 in Rat samples. It is validated for Western Blot.Spécification
| IL-1Rrp2/IL-1R6 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-1 R6, IL-1 Rrp2, IL1RL2, IL-1RL2, IL1Rrp2, IL-1Rrp2, IL1RRP2IL1R-rp2Interleukin-1 receptor-related protein 2, interleukin 1 receptor-like 2, interleukin-1 receptor-like 2 | |
| Il1rl2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 131011 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Rat | |
| Q62929 | |
| 8808 | |
| Mouse myeloma cell line NS0-derived recombinant rat IL-1 Rrp2/IL-1 R6 Gly22-Tyr340 Accession # Q62929 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rat IL-1 Rrp2/IL-1 R6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human (rh) IL-1 Rrp2 is observed and no cross-reactivity with recombinant mouse (rm) IL-1 RI, rmIL-1 RII, rhIL-1 RAcP, rmIL-1 R5, rmIL-1 R7, and rmIL-1 RAPL1 is observed. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit