missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IL-1Rrp2/IL-1R6 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB573
This item is not returnable.
View return policy
Description
IL-1Rrp2/IL-1R6 Monoclonal specifically detects IL-1Rrp2/IL-1R6 in Rat samples. It is validated for Western Blot.
Specifications
| IL-1Rrp2/IL-1R6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-1 R6, IL-1 Rrp2, IL1RL2, IL-1RL2, IL1Rrp2, IL-1Rrp2, IL1RRP2IL1R-rp2Interleukin-1 receptor-related protein 2, interleukin 1 receptor-like 2, interleukin-1 receptor-like 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8808 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 131011 | |
| Western Blot 1 ug/mL | |
| Q62929 | |
| Il1rl2 | |
| Mouse myeloma cell line NS0-derived recombinant rat IL-1 Rrp2/IL-1 R6 Gly22-Tyr340 Accession # Q62929 | |
| 500 μg | |
| Primary | |
| Detects rat IL-1 Rrp2/IL-1 R6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human (rh) IL-1 Rrp2 is observed and no cross-reactivity with recombinant mouse (rm) IL-1 RI, rmIL-1 RII, rhIL-1 RAcP, rmIL-1 R5, rmIL-1 R7, and rmIL-1 RAPL1 is observed. | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction