missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 RI Antibody (10E2), Novus Biologicals™
Mouse Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | IL-1 RI |
|---|---|
| Clone | 10E2 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30036136
|
Novus Biologicals
NBP3-26202-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30037409
|
Novus Biologicals
NBP3-26202-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-1 RI Monoclonal antibody specifically detects IL-1 RI in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| IL-1 RI | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
| Recombinant Human IL-1 RI protein [UniProt P14778] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 10E2 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 3554 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title