missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 RI Antibody (10E2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26202-100ul
This item is not returnable.
View return policy
Description
IL-1 RI Monoclonal antibody specifically detects IL-1 RI in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| IL-1 RI | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 10E2 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
| Recombinant Human IL-1 RI protein [UniProt P14778] | |
| 100 μL | |
| Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
| 3554 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction