missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 beta/IL-1F2 Antibody (43N3D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£184.00 - £315.00
Specifications
| Antigen | IL-1 beta/IL-1F2 |
|---|---|
| Clone | 43N3D8 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1-5ug/ml, Flow Cytometry 0.5ug/10^6cells, Immunohistochemistry-Paraffin 5ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18785903
|
Novus Biologicals
NBP2-27345SS |
0.025 mg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18798753
|
Novus Biologicals
NBP2-27345 |
£315.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IL-1 beta/IL-1F2 | |
| 1.0 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Apoptosis, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| P01584 | |
| 3553 | |
| Full-length recombinant human Interleukin-1 beta protein was used as immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 43N3D8 | |
| Western Blot 1-5ug/ml, Flow Cytometry 0.5ug/10^6cells, Immunohistochemistry-Paraffin 5ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
| IL1B | |
| IgG1 κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title