missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 beta/IL-1F2 Antibody (43N3D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-27345SS
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-1 beta/IL-1F2 | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 1-5ug/ml, Flow Cytometry 0.5ug/10^6cells, Immunohistochemistry-Paraffin 5ug/ml | |
| catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 43N3D8 | |
| Unconjugated | |
| P01584 | |
| IL1B | |
| Full-length recombinant human Interleukin-1 beta protein was used as immunogen for this antibody. | |
| 0.025 mg | |
| Adaptive Immunity, Apoptosis, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 3553 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction