missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 alpha/IL-1F1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-11365-0.025mg
This item is not returnable.
View return policy
Description
IL-1 alpha/IL-1F1 Polyclonal antibody specifically detects IL-1 alpha/IL-1F1 in Porcine samples. It is validated for Western Blot, ELISA
Specifications
| IL-1 alpha/IL-1F1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Pig | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| Hematopoietin-1, IL1, IL-1 alpha, IL-1A, IL1-ALPHA, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
| Recombinant porcine IL-1 ALPHA | |
| 0.025 mg | |
| Apoptosis, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 3552 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction