missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 alpha/IL-1F1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£224.00 - £654.00
Specifications
| Antigen | IL-1 alpha/IL-1F1 |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18660099
|
Novus Biologicals
NBP3-11365-0.025mg |
0.025 mg |
£224.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18684348
|
Novus Biologicals
NBP3-11365-0.1mg |
0.1 mg |
£654.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-1 alpha/IL-1F1 Polyclonal antibody specifically detects IL-1 alpha/IL-1F1 in Porcine samples. It is validated for Western Blot, ELISASpecifications
| IL-1 alpha/IL-1F1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Pig | |
| Hematopoietin-1, IL1, IL-1 alpha, IL-1A, IL1-ALPHA, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
| Recombinant porcine IL-1 ALPHA | |
| Primary | |
| Store at 4°C. Do not freeze. |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| PBS | |
| 3552 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title