missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IKK gamma Monoclonal Antibody (K94032_19C6), Invitrogen™

Mouse Monoclonal Antibody

£453.00 - £760.00

Specifications

Antigen IKK gamma
Clone K94032_19C6
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30204758
View Documents
Invitrogen™
MA556243
200 μL
£760.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30206993
View Documents
Invitrogen™
MA555863
50 μL
£453.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence of this protein is as follows: KLDLKRQKEQ ALREVEHLKR CQQQMAEDKA SVKAQVTSLL GELQESQSRL EAATKECQAL EGRARAASEQ ARQLESEREA LQQQHSVQVD QLRMQGQSVE AALRMERQAA SEEKRKLAQL QVAYHQLFQE YDNHIKSSVV GSERKRGMQL EDLKQQLQQA EEALVAKQEV IDKLKEEAEQ HKIVMETVPV LKAQADIYKA DFQAERQARE KLAEKKELLQ EQLEQLQREY SKLKASCQES ARIEDMRKRH

IKK gamma Monoclonal antibody specifically detects IKK gamma in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Specifications

Specifications

IKK gamma
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
1110037D23Rik; AI848108; AI851264; AMCBX1; AW124339; FIP3; FIP-3; Fip3p; gikk; I-kappa-B kinase gamma; I-kappa-B kinase subunit gamma; ikB kinase subunit gamma; ikB kinase-associated protein 1; IKBKG; IKK[g]; IKK3; IKKAP1; IKKG; IKK-gamma; IMD33; incontinentia pigmenti; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma; inhibitor of kappaB kinase gamma; inhibitor of nuclear factor kappa-B kinase subunit gamma; IP; IP1; IP2; IPD2; mF; mFIP-3; Nemo; NF-kappa B essential modulator; NF-kappa-B essential modifier; NF-kappa-B essential modulator; OTTHUMP00000207079; OTTHUMP00000212188; ZC2HC9
Recombinant human IKBKG (immunogen range is 111-360).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Ikbkg
K94032_19C6
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Mouse
Human
Q9Y6K9
8517
IgG2a
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.