missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IKK gamma Monoclonal Antibody (K94032_19C6), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556243

 View more versions of this product

Product Code. 30204758

  • £760.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: KLDLKRQKEQ ALREVEHLKR CQQQMAEDKA SVKAQVTSLL GELQESQSRL EAATKECQAL EGRARAASEQ ARQLESEREA LQQQHSVQVD QLRMQGQSVE AALRMERQAA SEEKRKLAQL QVAYHQLFQE YDNHIKSSVV GSERKRGMQL EDLKQQLQQA EEALVAKQEV IDKLKEEAEQ HKIVMETVPV LKAQADIYKA DFQAERQARE KLAEKKELLQ EQLEQLQREY SKLKASCQES ARIEDMRKRH

IKK gamma is the regulatory subunit of the inhibitor of the kappaB kinase (IKK) complex, which activates NF-kappaB resulting in activation of genes involved in inflammation, immunity, cell survival, and other pathways. Mutations in this gene result in incontinentia pigmenti, hypohidrotic ectodermal dysplasia, and several other types of immunodeficiencies. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene highly similar to this locus is located in an adjacent region of the X chromosome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IKK gamma
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Q9Y6K9
Ikbkg
Recombinant human IKBKG (immunogen range is 111-360).
200 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
K94032_19C6
Unconjugated
Ikbkg
1110037D23Rik; AI848108; AI851264; AMCBX1; AW124339; FIP3; FIP-3; Fip3p; gikk; I-kappa-B kinase gamma; I-kappa-B kinase subunit gamma; ikB kinase subunit gamma; ikB kinase-associated protein 1; IKBKG; IKK[g]; IKK3; IKKAP1; IKKG; IKK-gamma; IMD33; incontinentia pigmenti; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma; inhibitor of kappaB kinase gamma; inhibitor of nuclear factor kappa-B kinase subunit gamma; IP; IP1; IP2; IPD2; mF; mFIP-3; Nemo; NF-kappa B essential modulator; NF-kappa-B essential modifier; NF-kappa-B essential modulator; OTTHUMP00000207079; OTTHUMP00000212188; ZC2HC9
Mouse
Protein A
RUO
8517
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.