missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKB zeta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £304.00
Specifications
| Antigen | IKB zeta |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-3 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18028365
|
Novus Biologicals
NB100-56390SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18039393
|
Novus Biologicals
NB100-56390 |
0.1 mg |
£304.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IKB zeta Polyclonal specifically detects IKB zeta in Mouse samples. It is validated for Western Blot.Specifications
| IKB zeta | |
| Western Blot 1-3 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| FLJ30225, FLJ34463, ikappaBzeta, I-kappa-B-zeta, IkappaB-zeta, ikbzeta, IkB-zeta, IL-1 inducible nuclear ankyrin-repeat protein, INAPIkappa B-zeta variant 3, MAILIKBZikB-zeta, Molecule possessing ankyrin repeats induced by lipopolysaccharide, NF-kappa-B inhibitor zeta, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
| NFKBIZ | |
| IgG | |
| Protein G purified | |
| This antibody is reactive against Mouse. Predicted cross-reactivity based on sequence identity: Mouse (100%), Human, (100%), Rat (100%), Chimp (100%), Bovine (100%), Chicken (93%). Not yet tested in other species. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Q9BYH8, Q9EST8 | |
| 64332 | |
| Synthetic peptides corresponding to an amino acid range between 270-320 and 670-720 of mouse IkB zeta protein were used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title